Learn More
Waters Corp Prep Column, ODS1 Phase, OBD Technology, 10 μm, 10 x 250 mm
Supplier:  Waters Corp 186008285
Encompass_Preferred
Spherisorb OBD Preparative Columns are produced with patented Optimum Bed Density (OBD) Technology. They are widely referenced in scientific literature, providing a range of validated methods and applications for method development.
  • Produced with patented Optimum Bed Density (OBD) Technology.
  • Widely referenced in scientific literature.
  • Over 20,000 analytical abstracts published using Spherisorb columns.
  • Provides a range of validated methods and applications for method development.
  • Available in various particle sizes and bonded phases.
  • Offers unique separation selectivity.
Catalog No. 50-239-1042