Learn More
Cell Biolabs IncSupplier Diversity Partner SPHINGOMYELIN ASSAY KIT  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA601
Sphingomyelin Assay Kit measures the sphingomyelin levels present within serum, plasma, or tissue samples. Results are read with a 96-well fluorometric plate reader. 96 assays including standards and unknowns.
Catalog No. 50-673-645