Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPICE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP170712
Description
SPICE1 Polyclonal specifically detects SPICE1 in Human samples. It is validated for Western Blot.Specifications
SPICE1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CCDC52, FLJ26064, SPICE, spindle and centriole associated protein 1 | |
Rabbit | |
96 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Mouse: 100%; Human: 100%; Rat: 92%; Sumatran orangutan: 92%; Bovine: 78%; Canine: 78%;. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8N0Z3 | |
SPICE1 | |
Synthetic peptides corresponding to CCDC52(coiled-coil domain containing 52) The peptide sequence was selected from the middle region of CCDC52 (NP_653319). Peptide sequence KEQNWEEKTLPIDTDIQNSSEENRLFTQRWRVSHMGEDLENKTQAPFVNL. | |
Affinity purified | |
RUO | |
152185 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction