Learn More
ACROBiosystems SARS coronavirus CUHK-W1 Spike NTD Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems SPDS52H9100UG
Encompass_Preferred
SARS coronavirus CUHK-W1 Spike NTD Protein, His Tag (SPD-S52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Lys 291 (Accession # AAP13567.1).
Catalog No. 50-233-1725