Learn More
NPS Corp. Spilfyter™ Aqualockit Solidifiers
Turns liquids into easy to clean gels
Manufacturer:  NPS Corp. 670001
 View more versions of this product
Catalog No. 19-066-536
Add to cart
Description
Description
  • Absorbent granular-like material
  • For effective clean-up of contaminated water, sludge, neutralized acids/bases, low-level radioactive water
  • Component: Potassium Polyacrylate
Order Info
Packaging options: 10 Shaker bottles (10kg/23 lb.); 5 Gal. Bucket (9kg/19 lb.)
Specifications
Specifications
For easy and effective clean-up of contaminated water, sludge, neutral liquids or bases or low-level radioactive water
10 Case
10.43kg
Safety
Polymer
23 lb.
Super Absorbent Solidifier Shaker Bottle