Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPS Corp. Spilfyter™ KolorSafe™ Neutralizers for Acids

Take the guesswork out of neutralizing hazardous spills

Manufacturer:  NPS Corp. 410055

 View more versions of this product

Catalog No. 19-066-519

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.

  • Sodium-free liquid reagent changes color when neutralization is complete
  • Safe for use in laboratories and cleanrooms

Specifications

Specifications

55 gal.
55 gal., 208.2L
Liquid
Each
238kg
Safety
208.2L
For spill control
Color change indicator when neutralized
525 lb.
Chemical Neutralizer
Safety and Handling

Safety and Handling

ShelfLife : 3 Years

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit