Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FyterTech Nonwovens Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Take the guesswork out of neutralizing hazardous spills
Supplier: FyterTech Nonwovens 430004
Description
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
- Sodium-free liquid reagent changes color when neutralization is complete
- Safe for use in laboratories and cleanrooms
Specifications
1 gal. | |
Base Neutralizer | |
For lab and cleanrooms | |
4/Cs. | |
17 kg |
3.78 L | |
Liquid | |
Color change indicator when neutralized | |
38 lb. |
Safety and Handling
ShelfLife : 3 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction