Learn More
NPS Corp. Spilfyter™ Oil-Only Containment Products: Rolls
Use components together for optimum performance
Manufacturer:  NPS Corp. PS90
Catalog No. 19-066-615
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Fine fibers and state-of-the-art multilayering technology provide superior capillary action.
Compliance
Static-resistance sorbents meet NFPA specifications.
Specifications
Specifications
NFPA
144 ft. x 36 in. (43.8m x 91.4cm)
144 ft.
Spark
91.4cm
Safety
White
Absorbing flammable liquids such as jet fuel or xylene
43.8m
36 in.
Oil Only Static Resistant Sorbent