Learn More
FyterTech Nonwovens Spilfyter™ Oil-Only Containment Products: Truk Kits
Use components together for optimum performance
Supplier: FyterTech Nonwovens 311002
Description
Fine fibers and state-of-the-art multilayering technology provide superior capillary action.
Compliance
Static-resistance sorbents meet NFPA specifications.
Specifications
NFPA | |
49 L | |
Designed to fit behind the back seat of a truck | |
15 lb. |
13 gal. | |
Large Truck Kit, Oil-only | |
(3) 3 in. x 10 ft. socks (M-37), (15) 16 x 18 in. pads (M-75), (3) 18 x 18 in. pillows (M-62), 1 pair nitrile gloves, 1 pair splash goggles, disposal bag and twist tie | |
7 kg |