Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spilfyter™ Universal Spill Containment Products

Socks, pillows and drum tops for acids or bases, and nonagressive spills, such as oil, coolants, solvents, and water

Manufacturer:  NPS CorporationSupplier Diversity Partner G76

 View more versions of this product

Catalog No. 19-066-578


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Universal Sorbent Drum Top
41.58L
Black
55.88cm
Absorb and retain oil, water, solvents, coolants, etc., place on drum tops to soak up pump drips and keep work areas neat and clean
Precut hole, fine fiber sorbent
25 Case
3.17kg
11 gal.
11 gal., 41.58L
22 in.
N/A
55 gal. drums
Polypropylene
7 lb.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit