Learn More
Brady™ SpillFix™ Granular Absorbent Boom
Completely natural producing no carcinogens or dust and has more than four times absorption power of similar booms.
$68.80 - $78.10
Specifications
Product Type Coconut Coir Filled Boom
Type SOC
Diameter (Metric) Outer 10.1cm
Color Yellow
Material Lint-Free, Non-Abrasive
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Packaging Quantity Price Quantity & Availability  
19-666-500 View Documents Brady™
SF10BOOM
3.4m 2/Cs. Case of 2 for $68.80
Add to cart
19-666-501 View Documents Brady™
SF5BOOM
1.5m 4/Cs. Case of 4 for $78.10
Add to cart
Description
Description
  • Effective on absorption of hydrocarbons, water-based fluids and most aggressive chemicals
  • Certified safe for landfill disposal
  • Lightweight and easy to deploy
  • Wringable for added life
Specifications
Specifications
Coconut Coir Filled Boom
10.1cm
Lint-Free, Non-Abrasive
SOC
Yellow
22.7L (6 gal.)