Learn More
Justrite™ SpillSlope™ Steel Shelf For Under Fume Hood Safety Cabinet
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf For Under Fume Hood Safety Cabinet adds additional storage to cabinets.
$165.40 - $185.30
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
19-822-484
View Documents
Justrite™
29950
15 gal. Under Fume Hood cabinets
Each for $165.40
Add to Cart
19-822-485
View Documents
Justrite™
29951
19 gal. Under Fume Hood cabinets
Each for $165.40
Add to Cart
19-822-486
View Documents
Justrite™
29949
23 gal. Under Fume Hood cabinets
Each for $185.30
Add to Cart
19-822-487
View Documents
Justrite™
29953
31 gal. Under Fume Hood cabinets
Each for $183.10
Add to Cart
Description
Description
  • Protect workers, Reduce fire risks, and improve productivity by storing flammable fuels and chemicals in code-compliant safety cabinets
  • Designed to meet OSHA and NFPA standards, Sure-Grip EX cabinets are constructed of sturdy 18-gauge thick double-wall welded steel with 1-1/2 in. of insulating air space for fire resistance
  • High performance, self-latching doors close easily and securely for maximum protection under fire conditions
  • Fail-safe closing mechanism ensures three-point stainless steel bullet latching system works every time
  • For added security, U-Loc handle comes with a cylinder lock and key set or accepts an optional padlock
  • Haz-Alert reflective warning labels are highly visible under fire conditions or during power outages when illuminated with a flashlight
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof bottom sump
  • They adjust on 3 in. centers for versatile storage
  • Cabinets also feature dual vents with flame arresters, four adjustable self-leveling feet, grounding connector, and trilingual warning label
  • Durable lead-free epoxy/polyester powder-coat finish provides excellent chemical resistance
  • The ChemCor thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals
  • Seam-free surface eliminates the need for rivets or fasteners which risk corrosion from seep-through chemicals
  • Non-porous surface resists staining and cleans up easily