Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Spin-X™ UF Concentrators Available on GSA/VA Contract for Federal Government customers only.

Disposable, single use only ultrafiltration devices with polyethersulfone membranes (PES) for the centrifugal concentration and/or purification of biological fluids and aqueous solutions

Manufacturer:  Corning™ 431488

 View more versions of this product

Catalog No. 07-201-351


Add to cart

Description

Description

Providing that the appropriate device size and membrane cut-off is selected, Corning Spin-X UF products will typically yield recoveries of the concentrated sample in excess of 90% when the starting sample contains over 0.1mg/mL of the solute of interest. Depending on sample characteristics relative to the membrane type used, solute adsorption
on the membrane surface is typically 2 to 10 ++g/cm2. This can increase to 20 to 100 ++g/cm2
when the filtrate is of interest and the solute must pass through the whole internal structure
of the membrane.

  • General purpose polyethersulfone membranes for excellent retentate recovery
  • Llow fouling, exceptional flux and broad pH range Polyethersulfone membranes exhibit no hydrophobic or hydrophilic interactions
  • Advanced designs and low adsorption materials offer faster processing speeds and higher recovery of the concentrated samples

Corning offers Spin-X™ UF concentrators in three sizes.
Spin-X™ UF 500 for 100 to 500µL samples

  • Made of Polycarbonate body and concentrator cap; Polypropylene Filtrate vessel
  • One step procedure for sample preparation
  • Effectively be used in fixed angle rotors accepting 2.2mL centrifuge tubes
  • Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations, even with particle laden solutions
Spin-X™ UF 6 for 2 to 6mL samples
  • Made of Polycarbonate body and Filtrate vessel; Polypropylene concentrator cap
  • Offer increased volume flexibility and performance
  • Process up to 6mL in swing bucket or fixed angle rotors accepting standard 15mL conical bottom tubes
  • In a single spin, solutions can be concentrated in excess of 100-fold
  • Samples are typically concentrated in 10 to 30 minutes with macromolecular recoveries in excess of 95%
  • Features twin vertical membranes for unparalleled filtration speeds and 100x plus concentrations
  • Remaining volume is easy to read off the printed scale on the side of the concentrator and the modified dead stop pocket further simplifies direct pipet recovery of the final concentrate
Spin-X™ UF 20 for 5 to 20mL samples
  • Made of Polycarbonate body and Filtrate vessel; Polypropylene concentrator cap
  • Offer increased volume flexibility and performance
  • Handles up to 20mL in swing bucket centrifuges and 14mL in 25° fixed angle rotors accepting 50mL centrifuge tubes
  • Featuring twin vertical membranes for unparalleled filtration speeds and achieve 100x plus concentrations
  • Remaining volume is easy to read off the printed scale on the side of the concentrator and the modified dead stop pocket further simplifies direct pipet recovery of the final concentrate

General purpose laboratory concentration and desalting of proteins, enzymes, cells, biomolecules, antibodies and immunoglobulins
Removal of labeled amino acids and nucleotides
HPLC sample preparation
Deproteinization of samples
Recovery of biomolecules from cell culture supernatants, lysates

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Polypropylene
1.18 in.
Polyethersulfone
6cm2
4.56 in.
10000 MWCO
14mL
Ultrafiltration Concentrator
20mL
White
3cm
Centrifuge
Swinging bucket or fixed angle rotors
11.6cm
0.47 oz.
Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins, removal of labeled amino acids and nucleotides, HPLC sample preparation, deproteinization of samples, recovery of biomolecules from cell culture supernatants, lysates
12 Pack
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit