Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPIN4 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS17668R
Description
This gene is a member of the SPINDLIN family. SPINDLIN was first reported as an abundant maternal transcript present in the unfertilized egg and 2-cell, but not the 8-cell, stage embryo during the transition from oocyte to embryo in the mouse. This family is involved in gametogenesis, and the expression of genes in this family can be detected in early embryos.Specifications
SPIN4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SPIN4 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
139886 |
SPIN4 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q56A73 | |
FLJ44984; MGC133224; SPIN4; spindlin family member 4; spindlin family, member 4; Spindlin-4; TDRD28 | |
KLH conjugated synthetic peptide derived from human SPIN4. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title