Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Spinasterol, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1mg
This item is not returnable. View return policy
481-18-5
C29H48O
412.702
JZVFJDZBLUFKCA-FXIAWGAOSA-N
spinasterol,alpha-spinasterol,bessisterol,condrillasterol,hitodesterol,chondrillasterol,unii-0lg993qx1a,alpha-spinasterin,stigmasta-7,22e-dien-3beta-ol,5-alpha-stigmasta-7,22-dien-3-beta-ol, e
5281331
CHEBI:10333
(3S,5S,9R,10S,13R,14R,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,5,6,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
CCC(C=CC(C)C1CCC2C1(CCC3C2=CCC4C3(CCC(C4)O)C)C)C(C)C
This item is not returnable. View return policy
481-18-5
C29H48O
412.702
JZVFJDZBLUFKCA-FXIAWGAOSA-N
spinasterol,alpha-spinasterol,bessisterol,condrillasterol,hitodesterol,chondrillasterol,unii-0lg993qx1a,alpha-spinasterin,stigmasta-7,22e-dien-3beta-ol,5-alpha-stigmasta-7,22-dien-3-beta-ol, e
5281331
CHEBI:10333
(3S,5S,9R,10S,13R,14R,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,5,6,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
CCC(C=CC(C)C1CCC2C1(CCC3C2=CCC4C3(CCC(C4)O)C)C)C(C)C

TRPV1 antagonist; active in vivo

TRPV1 antagonist (IC50 = 1.4 μM). Reduces capsaicin-mediated Ca2+ influx in vitro; displays antinociceptive and antiedematogenic effects in multiple mouse models of pain. Brain penetrant and orally bioavailable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 1mg
Formula Weight 412.69
Percent Purity >95%
Chemical Name or Material α-Spinasterol

Recommended Storage : Store at -20°C