Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINK4 Rabbit anti-Human, DyLight 680, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP299857FR
Description
SPINK4 Polyclonal antibody specifically detects SPINK4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SPINK4 | |
DyLight 680 | |
50mM Sodium Borate | |
gastrointestinal peptide, MGC133107, serine peptidase inhibitor, Kazal type 4, serine protease inhibitor Kazal-type 4, serine protease inhibitor, Kazal type 4 | |
Produced in rabbits immunized with purified, recombinant SPINK4 (NP_055286.1; Met 1-Cys 86). | |
Affinity purified | |
RUO | |
Primary | |
27290 |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title