Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINK4 Recombinant Rabbit Monoclonal Antibody (265), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA543142
This item is not returnable.
View return policy
Description
Specificity: Human SPINK4
SPINK4 Recombinant Monoclonal antibody specifically detects SPINK4 in Human samples. It is validated for ELISA.Specifications
SPINK4 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O60575 | |
SPINK4 | |
Recombinant Human SPINK4 Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
265 | |
Unconjugated | |
SPINK4 | |
epididymis luminal protein 136; gastrointestinal peptide; HEL136; PEC60; PEC-60; Peptide PEC-60 homolog; serine peptidase inhibitor, Kazal type 4; serine protease inhibitor Kazal-type 4; serine protease inhibitor, Kazal type 4; SPINK4 | |
Rabbit | |
Protein A | |
RUO | |
27290 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction