Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINK5 Monoclonal Antibody (C10), Invitrogen™
Mouse Monoclonal Antibody
$211.50 - $451.50
Specifications
Antigen | SPINK5 |
---|---|
Clone | C10 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543567
|
Thermo Scientific
MA543567 |
20 μL |
Each for $211.50
|
|
|||||
PIMA543566
|
Thermo Scientific
MA543566 |
100 μL |
Each for $451.50
|
|
|||||
Description
SPINK5 Monoclonal antibody specifically detects SPINK5 in Human samples. It is validated for Western Blot.Specifications
SPINK5 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
2310065D10Rik; AV238912; Hemofiltrate peptide HF6478; Hemofiltrate peptide HF7665; ISK5; LEKT1; LEKTI; LETK1; LETKI; lympho-epithelial Kazal-type-related inhibitor; lymphoepithelial Kazal-type-related inhibitor; Netherton syndrome; NETS; NS; serine peptidase inhibitor, Kazal type 5; serine protease inhibitor Kazal-type 5; serine protease inhibitor, Kazal type 5; SPINK5; VAKT1; VAKTI | |
SPINK5 | |
IgG1 κ | |
Protein A/G | |
Antibody |
C10 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9NQ38 | |
11005 | |
Gly699-Ser976 (Accession Q9NQ38), with N-terminal His Tag | |
Primary | |
-20°C | |
SPINK5 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title