Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPIRE2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SPIRE2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179813
|
Novus Biologicals
NBP179813 |
100 μL |
Each of 1 for $436.00
|
|
Description
SPIRE2 Polyclonal specifically detects SPIRE2 in Human samples. It is validated for Western Blot.Specifications
SPIRE2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
KIAA1832, MGC117166, protein spire homolog 2, Spir-2, SPIR2, spire homolog 2 (Drosophila) | |
SPIRE2 | |
IgG | |
Affinity Purified | |
80 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_115827 | |
84501 | |
Synthetic peptide directed towards the N terminal of human SPIRE2The immunogen for this antibody is SPIRE2. Peptide sequence EAQTVQSLGFAIYRALDWGLDESEERELSPQLERLIDLMANNDSEDSGCG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title