Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spironolactone, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
10 g
Description
Spironolactone used as potassium sparing diuretic. Also used primarily to treat ascites in patients with liver disease, low-renin hypertension, hypokalemia, and Conn’s syndrome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in chloroform and methanol. Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Soluble in chloroform and methanol. Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Type | Spironolactone |
Physical Form | Powder |
Melting Point | 207°C to 208°C |
Merck Index | 14,8760 |
Solubility Information | Soluble in chloroform and methanol. Slightly soluble in water. |
Formula Weight | 416.58 |
Color | White |
Quantity | 10 g |
Chemical Name or Material | Spironolactone |
Safety and Handling
Hazard Category | H351-H360F-H373 |
Hazard Statement | GHS H Statement H360-H302 May damage fertility or the unborn child. Harmful if swallowed. |
Precautionary Statement | P201-P202-P260-P281-P308+P313-P501c |
EINECSNumber | 200-133-6 |
RTECSNumber | TU4725000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction