Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spiroxatrine, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1054-88-2
C22H25N3O3
379.46
JVGBTTIJPBFLTE-UHFFFAOYSA-N
spiroxatrine,spiroxatrinum,spiroxatrine inn,spiroxamide,spiroxatrine r 5188,spiroxatrinum inn-latin,espiroxatrina inn-spanish,dsstox_cid_25198,dsstox_rid_80744,dsstox_gsid_45198
5268
8-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one
C1CN(CCC12C(=O)NCN2C3=CC=CC=C3)CC4COC5=CC=CC=C5O4
This item is not returnable. View return policy
1054-88-2
C22H25N3O3
379.46
JVGBTTIJPBFLTE-UHFFFAOYSA-N
spiroxatrine,spiroxatrinum,spiroxatrine inn,spiroxamide,spiroxatrine r 5188,spiroxatrinum inn-latin,espiroxatrina inn-spanish,dsstox_cid_25198,dsstox_rid_80744,dsstox_gsid_45198
5268
8-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one
C1CN(CCC12C(=O)NCN2C3=CC=CC=C3)CC4COC5=CC=CC=C5O4

Potent α2C antagonist

5-HT1A antagonist. More active and selective than spiperone. Also a very potent α2C adrenergic receptor antagonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 379.46
Percent Purity >98%
Chemical Name or Material Spiroxatrine

Recommended Storage : Store at Room Temperature