Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spiroxatrine, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1054-88-2
C22H25N3O3
379.46
JVGBTTIJPBFLTE-UHFFFAOYSA-N
spiroxatrine,spiroxatrinum,spiroxatrine inn,spiroxamide,spiroxatrine r 5188,spiroxatrinum inn-latin,espiroxatrina inn-spanish,dsstox_cid_25198,dsstox_rid_80744,dsstox_gsid_45198
5268
8-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one
C1CN(CCC12C(=O)NCN2C3=CC=CC=C3)CC4COC5=CC=CC=C5O4
This item is not returnable. View return policy
1054-88-2
C22H25N3O3
379.46
JVGBTTIJPBFLTE-UHFFFAOYSA-N
spiroxatrine,spiroxatrinum,spiroxatrine inn,spiroxamide,spiroxatrine r 5188,spiroxatrinum inn-latin,espiroxatrina inn-spanish,dsstox_cid_25198,dsstox_rid_80744,dsstox_gsid_45198
5268
8-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one
C1CN(CCC12C(=O)NCN2C3=CC=CC=C3)CC4COC5=CC=CC=C5O4

Potent α2C antagonist

5-HT1A antagonist. More active and selective than spiperone. Also a very potent α2C adrenergic receptor antagonist.
Quantity 50mg
Formula Weight 379.46
Percent Purity >98%
Chemical Name or Material Spiroxatrine

Recommended Storage : Store at Room Temperature