Learn More
WASP Spitfire Extruder with Spitfire Core HT
SCI_ED
The Spitfire Extruder with Spitfire Core HT is an extractable cartridge without Teflon for printing at high temperature using Spitfire Extruders.
Manufacturer:  WASP WU7016
 View more versions of this product
Catalog No. S4131956
Add to cart
Alert:
Small Parts - Not recommended for children under 3 years old.
Description
Description
  • Extractable cartridge for Spitfire Extruders without Teflon
  • For printing at high temperature
  • Suggested for ABS, PA, PC, HIPS, PA+C, ASA, PMMA, and PP
Specifications
Specifications
Spitfire Extruders
3D Printer Accessory
0.7 mm