Learn More
WASP Spitfire Extruder with Spitfire Core HT Products for Science Education
The Spitfire Extruder with Spitfire Core HT is an extractable cartridge without Teflon for printing at high temperature using Spitfire Extruders.
$40.00
Specifications
Product Type 3D Printer Accessory
For Use With (Equipment) Spitfire Extruders
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 3
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
S4131955 Products for Science Education
View Documents
WASP
WU7015
0.4 mm
Each for $40.00
Add to cart
S4131956 Products for Science Education
View Documents
WASP
WU7016
0.7 mm
Each for $40.00
Add to cart
S4131957 Products for Science Education
View Documents
WASP
WU7017
1.2 mm
Each for $40.00
Add to cart
Description
Description
  • Extractable cartridge for Spitfire Extruders without Teflon
  • For printing at high temperature
  • Suggested for ABS, PA, PC, HIPS, PA+C, ASA, PMMA, and PP
Specifications
Specifications
3D Printer Accessory
Spitfire Extruders