Learn More
WASP Spitfire Extruder with Spitfire Core LT Products for Science Education
The Spitfire Extruder with Spitfire Core LT is an extractable steel cartridge with Teflon used for printing at low temperature using Spitfire Extruders.
$40.00
Specifications
Product Type 3D Printer Accessory
Material Steel
For Use With (Equipment) Spitfire Extruders
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 3
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
S4131952 Products for Science Education
View Documents
WASP
WU7009
0.4 mm
Each for $40.00
Add to cart
S4131953 Products for Science Education
View Documents
WASP
WU7010
0.7 mm
Each for $40.00
Add to cart
S4131954 Products for Science Education
View Documents
WASP
WU7011
1.2 mm
Each for $40.00
Add to cart
Description
Description
Expand your printing possibilities with additional interchangeable cartridges and nozzle diameters with the simple turn of a screw.
  • Extractable steel cartridge with Teflon for WASP Spitfire Extruders
  • Single extruder for FDM printers
  • For printing at low temperature
  • Indicated for PLA, PETG, and FLEX
  • Fast and accurate printing
Specifications
Specifications
3D Printer Accessory
Steel
Spitfire Extruders