Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SPL 334

S-Nitrosoglutathione reductase inhibitor

$202.00 - $848.50

Specifications

Chemical Name or Material 4-[[2-[[(2-Cyanophenyl)methyl]thio]-4-oxothieno[3,2-d]pyrimidin-3(4H)-yl]methyl]benzoic acid
Molecular Formula C22H15N3O3S2
CAS 688347-51-5
Product Type Target: Other Reductase Inhibitors
Formula Weight 433.5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-091-0 View Documents Tocris Bioscience™
5809/10
10mg Each for $202.00
Add to cart
 
58-095-0 View Documents Tocris Bioscience™
5809/50
50mg Each for $848.50
Add to cart
 
Description

Description

S-Nitrosoglutathione reductase inhibitor. Prevents weight loss, and attenuates lung inflammation and fibrosis in a bleomycin injury model of interstitial lung disease in mice.
Specifications

Specifications

4-[[2-[[(2-Cyanophenyl)methyl]thio]-4-oxothieno[3,2-d]pyrimidin-3(4H)-yl]methyl]benzoic acid
688347-51-5
433.5
C22H15N3O3S2
Target: Other Reductase Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit