Learn More
Eisco™ Splash Head Traps - Pear Shaped Products for Science Education
For use in distillation assemblies between the flask and condenser to prevent the carry over of raw liquid
Manufacturer:  Eisco™ CH0852B
 View more versions of this product
Catalog No. S29483
Add to cart
Description
Description
  • Pear shaped splash head trap
  • For use between flask and condenser to prevent carry over of raw liquids
  • Made from high quality borosilicate 3.3 glass
  • Autoclavable
  • Specifications
    Specifications
    Distillation
    Borosilicate Glass 3.3
    Cone: 24/29
    Pear Shaped
    1
    19/26
    SDS
    Documents
    Provide Content Correction
    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Cancel Submit
    1 year limited manufacturer warranty