Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Regular Splinter Forceps, Sklar, 4-1/2 in., Sharp, Straight, Serrated, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21781
Econo Regular Splinter Forceps, Sklar, 4-1/2 in., Sharp, Straight, Serrated, Stainless Steel. Econo Splinter Forceps are multi-purpose thumb forceps with sharp tips used for removing small foreign objects. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 4-1/2 inches. It is sold in boxes of 12.
Catalog No. 50-143-7686