Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Splitomicin, Tocris Bioscience™

Catalog Number 154210
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
5690-03-9
C13H10O2
198.22
ISFPDBUKMJDAJH-UHFFFAOYSA-N
This item is not returnable. View return policy
5690-03-9
C13H10O2
198.22
ISFPDBUKMJDAJH-UHFFFAOYSA-N

Sir2p inhibitor

Inhibitor of Sir2p (IC50 = 60 μM), an NAD+-dependent Sir2 family deacetylase required for chromatin-dependent silencing in yeast.
Quantity 10 mg
Synonym 2-acetoxybenzonitrile, benzonitrile,2-acetyloxy, o-acetoxybenzonitrile, pubchem13326, 2-cyanophenyl acetate, #, acmc-1aso4, benzonitrile, 2-acetyloxy, acetic acid 2-cyanophenyl ester
SMILES O=C1CCC2=C3C=CC=CC3=CC=C2O1
IUPAC Name 1H,2H,3H-naphtho[2,1-b]pyran-3-one
PubChem CID 79791
Formula Weight 198.22
Percent Purity >97%
Chemical Name or Material Splitomicin
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit