Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPN Mouse anti-Human, Clone: 84-3C1, Abnova™

Mouse monoclonal antibody raised against native human SPN.

Manufacturer:  Abnova Corporation MAB13435

Catalog No. 89-934-953


Add to cart

Description

Description

Sialophorin (leukosialin) is a major sialoglycoprotein on the surface of human T lymphocytes, monocytes, granulocytes, and some B lymphocytes, which appears to be important for immune function and may be part of a physiologic ligand-receptor complex involved in T-cell activation.[supplied by OMIM]

Specifications

Specifications

SPN
84-3C1
Mouse monoclonal antibody raised against native human SPN.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
CD43/GPL115/LSN
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
SPN
Stimulated human leukocytes.
Protein A/G purification
RUO
Primary
6693
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit