Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPN Mouse anti-Human, Clone: TP1/36 (Biotin), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13919

Catalog No. 89-033-984


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human SPN.

  • High specificity
  • High reactivity
Specifications

Specifications

SPN
TP1/36
Mouse monoclonal antibody raised against native human SPN.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
SPN
Native purified SPN from myeloblastic KG1 cells.
Protein A/G purification
RUO
Primary
6693
Flow Cytometry
Biotin
Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
P16150
CD43|GPL115|LSN
Mouse
IgG1
100 ug
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)