Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPON1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116176
Description
SPON1 Polyclonal Antibody for Western Blot, IHC (P)
SPON1 is a cell adhesion protein that promotes the attachment of spinal cord and sensory neuron cells and the out growth of neurites in vitro. SPON1 may contribute to the growth and guidance of axons in both the spinal cord and the PNS. Is known to be a major factor for vascular smooth muscle cells.
Specifications
SPON1 | |
Polyclonal | |
Unconjugated | |
SPON1 | |
AI666765; AW455831; BC020531; D330035F22Rik; FSP; f-spondin; KIAA0762; MGC10724; Spon1; spondin 1; spondin 1, (f-spondin) extracellular matrix protein; spondin 1, extracellular matrix protein; spondin-1; Sponf; Vascular smooth muscle cell growth-promoting factor; VSGP; VSGP/F-spondin | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10418, 233744 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8VCC9, Q9HCB6 | |
SPON1 | |
KLH conjugated synthetic peptide between 15-43 amino acids from the N-terminal region of human VSGP/F-spondin. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction