Learn More
Alfa Aesar™ Sponge Cobalt, A-8B46, promoted with Nickel and Chromium
Manufacturer:  Alfa Aesar™ 4487136
Quantity 500g
 View more versions of this product
Catalog No. AA4487136
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00239945
UN1378
Sponge Cobalt A-8B46, Promoted with Nickel and Chromium
Specifications
Slurry In Water
UN1378
Sponge Cobalt A-8B46, Promoted with Nickel and Chromium
MFCD00239945
500g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H251-H334-H351-H373-H317
Self-heating: may catch fire.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: METAL CATALYST, WETTED
TSCA : TSCA