Alfa Aesar™ Sponge Cobalt, A-8B46, promoted with Nickel and Chromium

Manufacturer: Alfa Aesar™ 4487122

Quantity 100g

 View more versions of this product

Catalog No. AA4487122

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00239945
    UN Number UN1378
    Chemical Name or Material Sponge Cobalt A-8B46, Promoted with Nickel and Chromium

    Specifications

    Physical Form Slurry In Water
    Health Hazard 2 GHS H Statement
    H251-H334-H351-H373-H317
    Self-heating: may catch fire.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00239945
    Health Hazard 1 Danger
    TSCA TSCA
    UN Number UN1378
    DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: METAL CATALYST, WETTED
    Quantity 100g
    Chemical Name or Material Sponge Cobalt A-8B46, Promoted with Nickel and Chromium

    GHS H Statement
    H251-H334-H351-H373-H317
    Self-heating: may catch fire.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit