Alfa Aesar™ Sponge Copper, A-3B00, nonpromoted

Manufacturer: Alfa Aesar™ 4487018

Quantity 50g

 View more versions of this product

Catalog No. AA4487018

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00239961
    UN Number UN1378
    Chemical Name or Material Sponge Copper A-3B00, Nonpromoted

    Specifications

    Physical Form Slurry In Water
    Health Hazard 3 GHS P Statement
    P280-P281-P308+P313-P405-P420-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Store away from other materials.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00239961
    Quantity 50g
    UN Number UN1378
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: METAL CATALYST, WETTED
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H251-H351-H361
    Self-heating: may catch fire.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Chemical Name or Material Sponge Copper A-3B00, Nonpromoted

    GHS H Statement
    H251-H351-H361
    Self-heating: may catch fire.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P280-P281-P308+P313-P405-P420-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Store away from other materials.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit