Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Spot Remover Pad, Sklar, 6 in. x 9 in. Instrument Spot Remover Pad For Use On OR Grade Satin Finish Instruments - Pack of 10. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101415
Instrument Spot Remover Pad, Sklar, 6 in. x 9 in. Instrument Spot Remover Pad For Use On OR Grade Satin Finish Instruments - Pack of 10. Sklar's Instrument Spot Remover is a pad of plastic material used to scrub spots off OR-grade, satin finish instruments before sterilization.
Catalog No. 50-143-6981