Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPR Mouse anti-Human, Clone: OTI3D6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501960

Catalog No. 50-167-3055


Add to cart

Description

Description

SPR encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
Specifications

Specifications

SPR
Monoclonal
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P35270
SDR38C1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6697
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3D6
Unconjugated
SPR
Liquid
SPR
Full length human recombit protein of human SPR produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit