Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPR Mouse anti-Human, Non-human primate, Clone: OTI1F4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525309

Catalog No. PIMA525309


Add to cart

Description

Description

SPR encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
Specifications

Specifications

SPR
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P35270
SDR38C1, sepiapterin reductase, short chain dehydrogenase/reductase family 38C, member 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6697, 705317
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1F4
Unconjugated
SPR
Liquid
SPR
Full length human recombit protein of SPR produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit