Learn More
ORS Nasco Sprayon Citrus Cleaner/Degreaser
An outstanding biodegradable heavy-duty, multi-purpose degreaser and graffiti remover
Manufacturer: ORS Nasco 425S00757
Description
- Non-chlorinated formula has the cleaning power of solvents to remove grime, dirt, grease, and oil
- Ideal as a cleaner prior to painting
- Pleasant citrus scent
- No CFCs or HCFCs
- Dielectric strength: 40,400 volts
- Applications include walls and fixtures in restrooms, locker rooms, schools, public buildings, transit system vehicles and buildings; also for cleaning residue on molds, aluminum and concrete floors
Certifications
NSF reg. no. 127526; C1 rated.
Specifications
<-18 to 162°C | |
NSF 127526, C1 | |
16 oz. | |
Ideal as a cleaner prior to painting, walls and fixtures in restrooms, locker rooms, schools, public buildings, transit system vehicles and buildings, also for cleaning residue on molds, aluminum and concrete floors |