Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPRED3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520626
Description
A suggested positive control is human brain tissue lysate. PA5-20626 can be used with blocking peptide PEP-0746.
SPRED3 is a member of the Sprouty (see SPRY1; MIM 602465)/SPRED family of proteins that regulate growth factor-induced activation of the MAP kinase cascade (see MAPK1; MIM 176948).
Specifications
SPRED3 | |
Polyclonal | |
Unconjugated | |
SPRED3 | |
AW047143; D130060H24Rik; Eve-3; SPRED3; spred-3; sprouty related EVH1 domain containing 3; sprouty related, EVH1 domain containing 3; Sprouty-related EVH1 domain-containing protein; sprouty-related, EVH1 domain containing 3; sprouty-related, EVH1 domain-containing protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
399473 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q2MJR0 | |
SPRED3 | |
A 16 amino acid peptide near the center of the human Spred3. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction