Learn More
American Educational Products Spring Binding Knife Switches
Durable components for classroom use.
Specifications
For Use With (Equipment) | Bell or stranded wire |
---|---|
Material | Bakelite™ Plastic |
Catalog Number | Mfr. No. | Type | For Use With (Application) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Application) | Includes | Price | Quantity & Availability | |||
S78772
![]() |
American Educational Products
7S78772 |
Knife Switch | Bell or stranded wire to hook up electrical circuits without the need for banana cables | Single pole, double throw with three spring terminals |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S78771
![]() |
American Educational Products
7S78771 |
Spring Binding Post Lamp Holders and Switch | Classroom use | Single pole, single throw with two spring terminals |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Use with bell or stranded wire to hook up electrical circuits without the need for banana cables
- Switches formed in a Bakelite plastic
- Up to five wires can be held in place for the most elaborate circuits
Specifications
Bell or stranded wire | |
Bakelite™ Plastic |