Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPRN Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309442100UL
Description
SPRN Polyclonal specifically detects SPRN in Human samples. It is validated for Western Blot.Specifications
SPRN | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
bA108K14.1, FLJ41197, Protein shadoo, SHADOO, shadow of prion protein, shadow of prion protein homolog (zebrafish), SHOShadoo, Sprn | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human SPRN (NP_001012526). Peptide sequence AAAGLAAGSGWRRAAGPGERGLEDEEDGVPGGNGTGPGIYSYRAWTSGAG | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
503542 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction