Learn More
SPRY4 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 227651AP
Catalog No. 50-173-3940
Add to cart
Description
Description
SPRY4 is an inhibitor of the receptor-transduced mitogen-activated protein kinase (MAPK) signaling pathway. The protein is positioned upstream of RAS activation and impairs the formation of active GTP-RAS.
Specifications
Specifications
SPRY4
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9C004
Protein sprouty homolog 4, sprouty homolog 4 (Drosophila), Spry 4, SPRY4
Rabbit
IgG
150 μL
-20°C
Primary
81848
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPRY4
Liquid
SPRY4
SPRY4 Fusion Protein Ag18574
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human