Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPSB3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220480
Description
SPSB3 Polyclonal specifically detects SPSB3 in Human, Mouse samples. It is validated for Western Blot.Specifications
SPSB3 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
C16orf31, chromosome 16 open reading frame 31, splA/ryanodine receptor domain and SOCS box containing 3, SPRY domain-containing SOCS box protein SSB-3, SSB-3, SSB3SPRY domain-containing SOCS box protein 3 | |
Rabbit | |
39 kDa | |
0.1 mL | |
Signal Transduction | |
90864 | |
Human, Mouse | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q6PJ21 | |
SPSB3 | |
Recombinant protein encompassing a sequence within the center region of human SPSB3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction