Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPT16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP262799

Catalog No. NBP262799

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SPT16 Polyclonal antibody specifically detects SPT16 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SPT16
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Rabbit
IgG
50μg
Primary
Human
Western Blot
Western Blot 1:1000
Purified
Human SPT16 (Suppressor of Ty 16 homolog)
Protein G purified
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit