Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA19 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548509

Catalog No. PA548509

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May have a role in spermiogenesis.
Specifications

Specifications

SPATA19
Polyclonal
PBS with 0.09% sodium azide
Q7Z5L4, Q9DAQ9
Spermatogenesis-associated protein 19, mitochondrial, Spermatogenic cell-specific gene 1 protein, Spergen-1, SPATA19, SPERGEN1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
219938, 75469
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPATA19
Liquid
SPATA19
KLH conjugated synthetic peptide between 122-150 amino acids from the C-terminal region of human SPT19.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit