Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPT3 Antibody (OTI1A5), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274348JF549
Description
SPT3 Monoclonal specifically detects SPT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SPT3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SUPT3H | |
Full length recombinant protein of human SPT3 (NP_852001) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A5 | |
Western Blot, Immunohistochemistry | |
SPT3-like protein, SPT3SPT3L, suppressor of Ty (S.cerevisiae) 3 homolog, suppressor of Ty 3 homolog (S. cerevisiae), transcription initiation protein SPT3 homolog | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
8464.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction