Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTBN2 Mouse anti-Human, Mouse, Rat, Clone: SPTBN2/1247, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14991

Catalog No. 89-035-011

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mouse monoclonal antibody raised against partial recombinant human SPTBN2.

  • High specificity
  • High reactivity
Specifications

Specifications

SPTBN2
SPTBN2/1247
Mouse monoclonal antibody raised against partial recombinant human SPTBN2.
In 10 mM PBS.
Liquid
SPTBN2
Recombinant protein corresponding to amino acids 356-475 of human SPTBN2.
Protein A/G purification
RUO
Primary
6712
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined
O15020
GTRAP41|SCA5
Mouse
IgG3
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)