Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTBN2 Rabbit anti-Human, Unconjugated, Clone: SPTBN2/2894R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00634

Catalog No. 89-339-649


Add to cart

Description

Description

SPTBN2 Monoclonal antibody specifically detects SPTBN2 in Human samples. It is validated for WB-Ce, IHC-P, Flow Cyt.
Specifications

Specifications

SPTBN2
SPTBN2/2894R
spectrin, beta, non-erythrocytic 2
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
Unconjugated
SPTBN2
GTRAP41, SCA5
Recombinant protein corresponding to amino acids 356-475 of human SPTBN2
100 μg
Primary
6712
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit