Learn More
SPTBN4 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S393-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259321V
Catalog No. NBP259321V
Add to cart
Description
Description
SPTBN4 Monoclonal antibody specifically detects SPTBN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SPTBN4
S393-2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SPTBN4
Fusion protein amino acids 1621-1832 (C-terminal repeats 14 to 15) of human Beta4-spectrin
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects >200kDa. Does not cross react with other Beta-spectrins
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
beta-IV spectrin, KIAA1642, QV, spectrin beta chain, brain 3, spectrin, beta, non-erythrocytic 4, spectrin, non-erythroid beta chain 3, SPNB4, SPTBN3
Mouse
IgG1
100 ul
Primary
57731
Human, Mouse, Rat